4-Nitrophenyl hexanoate ≥98.0%
Leverantör: TCI
Synonyms:
Hexanoic Acid 4-Nitrophenyl Ester
H0484-1G
H0484-5G
TCIAH0484-1GEA
965
SEK
TCIAH0484-1G
TCIAH0484-5G
4-Nitrophenyl hexanoate ≥98.0%
4-Nitrophenyl hexanoate
Formel:
C₁₂H₁₅NO₄ MW: 237.26 g/mol Kokpunkt: 145°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00056111 CAS nummer: 956-75-2 |
Specifikation testresultat
Appearance | Light yellow to Brown clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...