Hexafluoroacetone trihydrate ≥97.0%
Leverantör: TCI
|
Fara
|
Synonyms:
Perfluoroacetone trihydrate
H0425-25G
H0425-5G
TCIAH0425-25GEA
855
SEK
TCIAH0425-25G
TCIAH0425-5G
Hexafluoroacetone trihydrate ≥97.0%
Hexafluoroacetone trihydrate
Formel:
C₃F₆O·3H₂O MW: 220.07 g/mol |
MDL Number:
MFCD00149061 CAS nummer: 34202-69-2 UN: 2552 ADR: 6.1 II Merck Index: 14,04685 |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (Neutralization titration) | min. 97.0 % (calcd.on anh.substance) |
Water | max. 25.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...