1-(2-Hydroxyethyl)-2-pyrrolidone ≥98.0%

Leverantör: TCI
Varning

Synonyms: N-(2-Hydroxyethyl)-2-pyrrolidone

H0408-25G H0408-500G
TCIAH0408-25GEA 215 SEK
TCIAH0408-25G TCIAH0408-500G
1-(2-Hydroxyethyl)-2-pyrrolidone ≥98.0%
1-(2-Hydroxyethyl)-2-pyrrolidone
Formel: C₆H₁₁NO₂
MW: 129.16 g/mol
Kokpunkt: 175°C
Flampunkt: 174°C
MDL Number: MFCD00014103
CAS nummer: 3445-11-2
EINECS: 222-359-4

Order Now

Specifikation testresultat

Appearance White or Colorles to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR