Heparin sodium salt (from hog intestine)

Leverantör: TCI EUROPE

Synonyms: Reviparin sodium

H0393-100MG H0393-1G
TCIAH0393-100MGEA 575 SEK
TCIAH0393-100MG TCIAH0393-1G
Heparin sodium salt (from hog intestine)
Heparin sodium salt
MDL Number: MFCD00081689
CAS nummer: 9041-08-1

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Total Nitrogen 1.3 to 2.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR