1-[N-(2-Hydroxyethyl)-4'-piperidyl]-3-(4'-piperidyl)propane ≥96.0% (by GC, titration analysis)
Leverantör: TCI
|
Varning
|
H0361-25G
H0361-5G
TCIAH0361-25GEA
4000
SEK
TCIAH0361-25G
TCIAH0361-5G
1-[N-(2-Hydroxyethyl)-4'-piperidyl]-3-(4'-piperidyl)propane ≥96.0% (by GC, titration analysis)
1-[N-(2-Hydroxyethyl)-4'-piperidyl]-3-(4'-piperidyl)propane
Formel:
C₁₅H₃₀N₂O Kokpunkt: 250°C |
MDL Number:
MFCD00023766 CAS nummer: 14712-23-3 EINECS: 238-761-8 |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 96.0 % |
Purity (Nonaqueous Titration) | min. 96.0 % |
Melting point | 82.0 to 86.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...