4-Hydroxysophthalic acid 98,0 HPLC, T

Leverantör: TCI
Varning

Synonyms: 4-Hydroxyisophthalic acid

H0257-25G H0257-5G
TCIAH0257-25GEA 3500 SEK
TCIAH0257-25G TCIAH0257-5G
4-Hydroxysophthalic acid 98,0 HPLC, T
4-Hydroxysophthalic acid
Formel: C₈H₆O₅
MW: 182.13 g/mol
Smältpunkt: 316
MDL Number: MFCD00010391
CAS nummer: 636-46-4
EINECS: 211-258-0
Merck Index: 14,04827
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=479bf5a200c9c9e95620e44266f4ced847217872&vid=b06b14ee1f9c9580d3e7afc0e7ac13bd587324f5&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR