Hinokitiol ≥99.0%

Leverantör: TCI EUROPE
Varning

Synonyms: beta-Thujaplicine, 2-Hydroxy-4-isopropylcyclohepta-2,4,6-trien-1-one

H0142-1G H0142-5G
TCIAH0142-1GEA 350 SEK
TCIAH0142-1G TCIAH0142-5G
Hinokitiol ≥99.0%
Hinokitiol
Formel: C₁₀H₁₂O₂
MW: 164,20 g/mol
Kokpunkt: 138°C
MDL Number: MFCD00059582
CAS nummer: 499-44-5
EINECS: 207-880-7
Merck Index: 14,09390

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Melting point 50.0 to 54.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR