Ginsenoside Rd ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
(2S,3R,4S,5S,6R)-2-(((2R,3R,4S,5S,6R)-4,5-Dihydroxy-2-(((3S,5R,8R,9R,10R,12R,13R,14R,17S)-12-hydroxy-4,4,8,10,14-pentamethyl-17-((S)-6-methyl-2-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hept-5-en-2-yl)hexadecahydro-1H-cyclopenta[a]phenanthren-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
G0550-100MG
G0550-25MG
TCIAG0550-100MGEA
4925
SEK
TCIAG0550-100MG
TCIAG0550-25MG
Ginsenoside Rd ≥98.0%
Ginsenoside Rd
Formel:
C₄₈H₈₂O₁₈ MW: 947,17 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00210508 CAS nummer: 52705-93-8 EINECS: 258-118-5 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) (CAD) | min. 98.0 area% |
Elemental analysis (Carbon) | 57.50 to 63.50 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...