N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-(3-morpholinopropoxy)quinazolin-4-amine ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Gefitinib
G0546-1G
G0546-5G
TCIAG0546-1GEA
430
SEK
TCIAG0546-1G
TCIAG0546-5G
N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-(3-morpholinopropoxy)quinazolin-4-amine ≥98.0% (by HPLC, titration analysis)
N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-(3-morpholinopropoxy)quinazolin-4-amine
Formel:
C₂₂H₂₄ClFN₄O₃ MW: 446,91 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD04307832 CAS nummer: 184475-35-2 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 194.0 to 198.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...