Gomisin K3 ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
Schisanhenol, 2,3,10,11,12-Pentamethoxy-6,7-dimethyl-5,6,7,8-tetrahydrodibenzo[a,c][8]annulen-1-ol
G0542-25MG
G0542-5MG
TCIAG0542-25MGEA
6650
SEK
TCIAG0542-25MG
TCIAG0542-5MG
Gomisin K3 ≥98.0%
Gomisin K3
Formel:
C₂₃H₃₀O₆ MW: 402,49 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD28411506 CAS nummer: 69363-14-0 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 128.0 to 132.0 deg-C |
Specific rotation [a]20/D | +70.0 to +77.0 deg (C=0.5, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...