Glycidyltrimethylammonium chloride ∼80% i vatten

Leverantör: TCI EUROPE
Fara

Synonyms: 2-Oxiranemethanaminium,N,N,N-trimethyl chloride, Glycidyltrimethylammoniumchloride, β,γ-Epoxypropyltrimethylammonium chloride, Trimethylglycidylammonium chloride

G0476-100G G0476-25G G0476-500G
TCIAG0476-100GEA 510 SEK
TCIAG0476-100G TCIAG0476-25G TCIAG0476-500G
Glycidyltrimethylammonium chloride ∼80% i vatten
Glycidyltrimethylammonium chloride
Formel: C₆H₁₄ClNO
MW: 151,635 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00011940
CAS nummer: 3033-77-0
EINECS: 221-221-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6934289fcf8c9fe0403ab1379ab36bd42fbf1e4&vid=a6934289fcf8c9fe0403ab1379ab36bd42fbf1e4&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Concentration (Nonaqueous Titration) 75.0 to 85.0 w/w%
Water 15.0 to 25.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR