[8-(Glycidyloxy)-n-octyl]trimethoxysilane ≥97.0%
Leverantör: TCI
|
Varning
|
G0469-5G
G0469-25G
TCIAG0469-5GEA
830
SEK
TCIAG0469-5G
TCIAG0469-25G
[8-(Glycidyloxy)-n-octyl]trimethoxysilane ≥97.0%
[8-(Glycidyloxy)-n-octyl]trimethoxysilane
Formel:
C₁₄H₃₀O₅Si |
MDL Number:
MFCD29472534 CAS nummer: 1239602-38-0 |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...