Genipin ≥97.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Methyl (1S,2R,6S)-2-Hydroxy-9-(hydroxymethyl)-3-oxabicyclo[4.3.0]nona-4,8-diene-5-carboxylate, (1R,4aS,7aS)-Methyl 1-hydroxy-7-(hydroxymethyl)-1,4a,5,7a-tetrahydrocyclopenta[c]pyran-4-carboxylate

G0458-25MG G0458-100MG
TCIAG0458-25MGEA 535 SEK
TCIAG0458-25MG TCIAG0458-100MG
Genipin ≥97.0%
Genipin
Formel: C₁₁H₁₄O₅
MW: 226,23 g/mol
MDL Number: MFCD00888600
CAS nummer: 6902-77-8
UN: 2811
ADR: 6.1,III

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 118.0 to 123.0 deg-C
Specific rotation [a]20/D +134 to +142 deg (C=1,MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR