Graphene, nanoplatelets, Length: 5 µm, Ø 6-8 nm
Leverantör: TCI
|
Varning
|
G0441-5G
G0441-25G
TCIAG0441-5GEA
750
SEK
TCIAG0441-5G
TCIAG0441-25G
Graphene, nanoplatelets, Length: 5 µm, Ø 6-8 nm
Graphene
MDL Number:
MFCD00144065 CAS nummer: 1034343-98-0 |
Specifikation testresultat
Appearance | Black powder to lump |
Thick | 6 to 8 nm |
Average size | Equal to 5 micro_m |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...