Geranyl tiglate ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
Tiglic acid Geranyl Ester, (E)-(E)-3,7-Dimethylocta-2,6-dien-1-yl 2-methylbut-2-enoate
G0430-5ML
G0430-25ML
TCIAG0430-5MLEA
245
SEK
TCIAG0430-5ML
TCIAG0430-25ML
Geranyl tiglate ≥98.0%
Geranyl tiglate
Formel:
C₁₅H₂₄O₂ MW: 236,36 g/mol Kokpunkt: 151°C |
MDL Number:
MFCD00016622 CAS nummer: 7785-33-3 EINECS: 232-078-9 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...