(S)-α-(2-Oxiranyl)anisole ≥98.0%
Leverantör: TCI
|
Fara
|
Synonyms:
(S)-Glycidyl phenyl ether
G0410-1G
G0410-5G
TCIAG0410-1GEA
415
SEK
TCIAG0410-1G
TCIAG0410-5G
(S)-α-(2-Oxiranyl)anisole ≥98.0%
(S)-α-(2-Oxiranyl)anisole
Formel:
C₉H₁₀O₂ MW: 150.1745 g/mol |
MDL Number:
MFCD06659026 CAS nummer: 71031-03-3 |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Specific rotation [a]20/D | +12.0 to +16.0 deg (C=1、EtOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...