Gestodene ≥98.0%

Leverantör: TCI
Fara

Synonyms: 13beta-Ethyl-17beta-hydroxy-18,19-dinorpregna-4,15-dien-20-yn-3-one

G0404-100MG G0404-1G
TCIAG0404-100MGEA 670 SEK
TCIAG0404-100MG TCIAG0404-1G
Gestodene ≥98.0%
Gestodene
Formel: C₂₁H₂₆O₂
Storage Temperature: Frys
MDL Number: MFCD00867858
CAS nummer: 60282-87-3
EINECS: 262-145-8
Merck Index: 14,04413
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df888f29f65b523e04d7e2a30b4657bece893d59&vid=df888f29f65b523e04d7e2a30b4657bece893d59&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 197.0 to 201.0 deg-C
Specific rotation [a]20/D -179 to -185 deg (C=0.5,CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR