(S)-N-Glycidylphthalimide ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
(S)-2-(Oxiran-2-ylmethyl)isoindoline-1,3-dione, (S)-N-(2,3-Epoxypropyl)phthalimide
G0328-25G
G0328-5G
TCIAG0328-25GEA
1675
SEK
TCIAG0328-25G
TCIAG0328-5G
(S)-N-Glycidylphthalimide ≥98.0%
(S)-N-Glycidylphthalimide
Formel:
C₁₁H₉NO₃ MW: 203,20 g/mol |
MDL Number:
MFCD04973350 CAS nummer: 161596-47-0 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 100.0 to 104.0 deg-C |
Specific rotation [a]20/D | 7.0 to 13.0 deg (C=2.2, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...