(R)-Glycidyl-3-nitrobenzenesulphonate ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
(R)-(-)-Glycidyl nosylate, 3-Nitrobenzenesulphonic acid (R)-Glycidyl Ester, (R)-Oxiran-2-ylmethyl 3-nitrobenzenesulphonate
G0286-25G
G0286-5G
TCIAG0286-25GEA
2025
SEK
TCIAG0286-25G
TCIAG0286-5G
(R)-Glycidyl-3-nitrobenzenesulphonate ≥98.0%
(R)-Glycidyl-3-nitrobenzenesulphonate
Formel:
C₉H₉NO₆S MW: 259,23 g/mol |
MDL Number:
MFCD00013436 CAS nummer: 115314-17-5 UN: 1325 ADR: 4.1,II |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 63.0 to 66.0 deg-C |
Specific rotation [a]20/D | -20.0 to -25.0 deg (C=1, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...