Magnesium(II) gluconate hydrate ≥98.0%

Leverantör: TCI EUROPE

Synonyms: D-Gluconic acid hemimagnesium salt hydrate, Magnesium D-gluconate hydrate, Gluconic acid magnesium(II) salt hydrate, D-Gluconic acid magnesium salt hydrate

G0276-25G G0276-500G
TCIAG0276-25GEA 215 SEK
TCIAG0276-25G TCIAG0276-500G
Magnesium(II) gluconate hydrate ≥98.0%
Magnesium(II) gluconate hydrate
Formel: C₆H₁₂O₇·xH₂O
MW: 414,60 g/mol (anhy)
MDL Number: MFCD00150971
CAS nummer: 3632-91-5
EINECS: 222-848-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cf5b3aecae7f7f9115340d091c64eaafeff8e52&vid=24c41bb0b4c747deb2954e925b890037ace85c1a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (Chelometric Titration) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D +10.0 to +13.0 deg (C=1, H2O)
Solubility in Water almost transparency
Water max. 13.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR