D-Glucal ≥97.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 1,5-Anhydro-2-deoxy-D-arabino-hex-1-enitol

G0274-1G G0274-5G
TCIAG0274-1GEA 870 SEK
TCIAG0274-1G TCIAG0274-5G
D-Glucal ≥97.0%
D-Glucal
Formel: C₆H₁₀O₄
MW: 146,14 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00067186
CAS nummer: 13265-84-4
EINECS: 236-259-3

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 57.0 to 62.0 deg-C
Specific rotation [a]20/D -7.0 to -12.0 deg (C=2, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR