D-Galactal ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 1,5-Anhydro-2-deoxy-D-lyxo-hex-1-enitol, (2R,3R,4R)-2-(Hydroxymethyl)-3,4-dihydro-2H-pyran-3,4-diol

G0273-1G G0273-5G
TCIAG0273-1GEA 845 SEK
TCIAG0273-1G TCIAG0273-5G
D-Galactal ≥98.0%
D-Galactal
Formel: C₆H₁₀O₄
MW: 146,14 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00038067
CAS nummer: 21193-75-9
EINECS: 244-264-7

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D -20.0 to -23.0 deg (C=1.2, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR