DL-Glyceraldehyde diethyl acetal ≥95.0%

Leverantör: TCI

G0216-25G G0216-5G
TCIAG0216-25GEA 7300 SEK
TCIAG0216-25G TCIAG0216-5G
DL-Glyceraldehyde diethyl acetal ≥95.0%
DL-Glyceraldehyde diethyl acetal
Formel: C₇H₁₆O₄
Kokpunkt: 136°C
MDL Number: MFCD00059599
CAS nummer: 10487-05-5

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR