Glutaconaldehydedianil hydrochloride ≥98.0%
Leverantör: TCI
|
Varning
|
G0201-25G
G0201-250G
TCIAG0201-25GEA
535
SEK
TCIAG0201-25G
TCIAG0201-250G
Glutaconaldehydedianil hydrochloride ≥98.0%
Glutaconaldehydedianil hydrochloride
Formel:
C₁₇H₁₆N₂ |
MDL Number:
MFCD00012630 CAS nummer: 1497-49-0 EINECS: 216-094-3 |
Specifikation testresultat
Appearance | Red to Dark red to Dark purple powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...