3-Allyloxy-1,2-propanediol ≥99.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Glycerol-1-allylether, Glycerol alpha-Monoallyl Ether

G0189-25G G0189-500G
TCIAG0189-25GEA 230 SEK
TCIAG0189-25G TCIAG0189-500G
3-Allyloxy-1,2-propanediol ≥99.0%
3-Allyloxy-1,2-propanediol
Formel: C₆H₁₂O₃
MW: 132,16 g/mol
Kokpunkt: 142°C
Flampunkt: 134°C
MDL Number: MFCD00004721
CAS nummer: 123-34-2
EINECS: 204-620-4

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR