Glycyl-l-glutamic acid

Leverantör: TCI

G0123-1G G0123-100MG
TCIAG0123-1GEA 1500 SEK
TCIAG0123-1G TCIAG0123-100MG
Glycyl-l-glutamic acid
Proteiner

Melting point: 158 °C
Specific rotation [a]20/D: –7° (C=5, H₂O)
Water: Max. 8,0%
Purity (nonaqueous titration): Min. 98,0% (calculated on anhydrous substance)


Product categories: Bioscience, Oligopeptides
Other product names/synonyms: H-Gly-Glu-OH

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR