DL-Glutamine ≥98.0%
Leverantör: TCI
Synonyms:
H-DL-Gln-OH, 2,5-Diamino-5-oxopentanoic acid
G0062-1G
G0062-10G
TCIAG0062-1GEA
470
SEK
TCIAG0062-1G
TCIAG0062-10G
DL-Glutamine ≥98.0%
DL-Glutamine
Formel:
C₅H₁₀N₂O₃ MW: 146.144 g/mol |
MDL Number:
MFCD00065103 CAS nummer: 6899-04-3 EINECS: 230-006-0 Merck Index: 14,04471 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...