4-Fluorophenethylamine hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: 2-(4-Fluorophenyl)ethanamine hydrochloride, 2-(4-Fluorophenyl)ethylammonium chloride, 4-Fluorophenethylamine hydrochloride, 2-(4-Fluorophenyl)ethylamine hydrochloride

F1256-1G F1256-5G
TCIAF1256-1GEA 535 SEK
TCIAF1256-1G TCIAF1256-5G
4-Fluorophenethylamine hydrochloride ≥98.0% (by HPLC, titration analysis)
4-Fluorophenethylamine hydrochloride
Formel: C₈H₁₁ClFN
MW: 175,63 g/mol
Smältpunkt: 208°C
MDL Number: MFCD00008190
CAS nummer: 459-19-8
EINECS: 207-285-2
UN: 2811
ADR: 6.1,III

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR