4-Fluoro-2-methoxy-5-nitroaniline ≥98.0%

Leverantör: TCI
Varning

F1246-25G F1246-5G
TCIAF1246-25GEA 1050 SEK
TCIAF1246-25G TCIAF1246-5G
4-Fluoro-2-methoxy-5-nitroaniline ≥98.0%
4-Fluoro-2-methoxy-5-nitroaniline
Formel: C₇H₇FN₂O₃
MDL Number: MFCD23098941
CAS nummer: 1075705-01-9

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 128.0 to 132.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR