Fluroxypyr 1-Methylheptyl ester ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1-Methylheptyl [(4-Amino-3,5-dichloro-6-fluoropyridin-2-yl)oxy]acetate, Fluroxypyr Meptyl
F1213-5G
F1213-25G
TCIAF1213-5GEA
645
SEK
TCIAF1213-5G
TCIAF1213-25G
Fluroxypyr 1-Methylheptyl ester ≥98.0% (by HPLC, total nitrogen)
Fluroxypyr 1-Methylheptyl ester
Formel:
C₁₅H₂₁Cl₂FN₂O₃ MW: 367,24 g/mol |
MDL Number:
MFCD00144318 CAS nummer: 81406-37-3 EINECS: 279-752-9 UN: 3077 ADR: 9,III Merck Index: 14,04203 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 56.0 to 60.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...