Fenspiride hydrochloride ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
8-Phenethyl-1-oxa-3,8-diazaspiro[4.5]decan-2-one hydrochloride, Fenspiride HCl
F1211-1G
F1211-5G
TCIAF1211-1GEA
430
SEK
TCIAF1211-1G
TCIAF1211-5G
Fenspiride hydrochloride ≥98.0% (by HPLC, titration analysis)
Fenspiride hydrochloride
Formel:
C₁₅H₂₀N₂O₂·HCl MW: 296,80 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00133315 CAS nummer: 5053-08-7 EINECS: 225-752-9 Merck Index: 14,03999 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...