8-Fluoroquinoline ≥97.0%
Leverantör: TCI EUROPE
Varning
F1185-1G F1185-5G
TCIAF1185-1GEA 295 SEK
TCIAF1185-1G TCIAF1185-5G
8-Fluoroquinoline ≥97.0%
8-Fluoroquinoline
Formel: C₉H₆FN
MW: 147,1500 g/mol
Kokpunkt: 148°C
MDL Number: MFCD01685514
CAS nummer: 394-68-3
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %