4-Fluoro-2-iodo-1-nitrobenzene ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
F1178-1G
F1178-5G
TCIAF1178-1GEA
695
SEK
TCIAF1178-1G
TCIAF1178-5G
4-Fluoro-2-iodo-1-nitrobenzene ≥98.0%
4-Fluoro-2-iodo-1-nitrobenzene
Formel:
C₆H₃FINO₂ MW: 267,00 g/mol Kokpunkt: 148°C Storage Temperature: Kylskåp |
MDL Number:
MFCD16658618 CAS nummer: 41860-64-4 |
Specifikation testresultat
Appearance | Light yellow to Brown powder to lump |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...