Formamidinium tetrafluoroborate ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
Formamidine tetrafluoroborate
F1152-1G
F1152-5G
TCIAF1152-1GEA
575
SEK
TCIAF1152-1G
TCIAF1152-5G
Formamidinium tetrafluoroborate ≥98.0%
Formamidinium tetrafluoroborate
Formel:
CH₅BF₄N₂ MW: 131,87 g/mol |
CAS nummer:
2607106-18-1 UN: 1759 ADR: 8,III |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...