3-Fluoro-5-(trifluoromethyl)benzyl bromide ≥98.0%
Leverantör: TCI EUROPE
Fara
F1131-1G F1131-5G
TCIAF1131-1GEA 375 SEK
TCIAF1131-1G TCIAF1131-5G
3-Fluoro-5-(trifluoromethyl)benzyl bromide ≥98.0%
3-Fluoro-5-(trifluoromethyl)benzyl bromide
Formel: C₈H₅BrF₄
MW: 257,03 g/mol
Kokpunkt: 43°C
MDL Number: MFCD00061175
CAS nummer: 239087-09-3
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2228f14e100e6cd1be98f1025982dfe074aa4c39&vid=2228f14e100e6cd1be98f1025982dfe074aa4c39&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %