4-Fluoro-2-(trifluoromethyl)benzenesulphonyl chloride ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
4-Fluoro-2-(trifluoromethyl)benzene-1-sulphonyl chloride
F1124-1G
F1124-5G
TCIAF1124-1GEA
415
SEK
TCIAF1124-1G
TCIAF1124-5G
4-Fluoro-2-(trifluoromethyl)benzenesulphonyl chloride ≥98.0%
4-Fluoro-2-(trifluoromethyl)benzenesulphonyl chloride
Formel:
C₇H₃ClF₄O₂S MW: 262,61 g/mol |
MDL Number:
MFCD01091000 CAS nummer: 176225-09-5 UN: 3261 ADR: 8,II |
Specifikation testresultat
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Melting point | 48.0 to 52.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...