5-(2-Fluorophenyl)pyrrole-3-carboxaldehyde ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
5-(2-Fluorophenyl)-1H-pyrrole-3-carbaldehyde
F1074-1G
F1074-5G
TCIAF1074-1GEA
670
SEK
TCIAF1074-1G
TCIAF1074-5G
5-(2-Fluorophenyl)pyrrole-3-carboxaldehyde ≥98.0%
5-(2-Fluorophenyl)pyrrole-3-carboxaldehyde
Formel:
C₁₁H₈FNO MW: 189,19 g/mol |
MDL Number:
MFCD11875867 CAS nummer: 881674-56-2 |
Specifikation testresultat
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 131.0 to 135.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...