4-Fluoro-3-formylphenylboronic acid (contains varying amounts of Anhydride)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-Fluoro-3-formylbenzeneboronic acid, (4-Fluoro-3-formylphenyl)boronic acid
F1061-1G
F1061-5G
TCIAF1061-1GEA
365
SEK
TCIAF1061-1G
TCIAF1061-5G
4-Fluoro-3-formylphenylboronic acid (contains varying amounts of Anhydride)
4-Fluoro-3-formylphenylboronic acid
Formel:
C₇H₆BFO₃ MW: 167,93 g/mol |
MDL Number:
MFCD02093074 CAS nummer: 374538-01-9 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 113.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...