5-Fluoro-2-methylindole ≥98.0%
Leverantör: TCI EUROPE
Varning
F1047-1G F1047-5G
TCIAF1047-1GEA 1350 SEK
TCIAF1047-1G TCIAF1047-5G
5-Fluoro-2-methylindole ≥98.0%
5-Fluoro-2-methylindole
Formel: C9H8FN
MW: 149,17 g/mol
Kokpunkt: 120°C
MDL Number: MFCD02093649
CAS nummer: 399-72-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6e06f3d450ec4028d8898feba1137ea3f824ab9&vid=d6e06f3d450ec4028d8898feba1137ea3f824ab9&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %