2-(4-Fluorophenyl)-4,6-diphenyl-1,3,5-triazine ≥98.0%

Leverantör: TCI EUROPE
Varning

F1045-200MG F1045-1G
TCIAF1045-200MGEA 375 SEK
TCIAF1045-200MG TCIAF1045-1G
2-(4-Fluorophenyl)-4,6-diphenyl-1,3,5-triazine ≥98.0%
2-(4-Fluorophenyl)-4,6-diphenyl-1,3,5-triazine
Formel: C₂₁H₁₄FN₃
MW: 327,36 g/mol
MDL Number: MFCD28964710
CAS nummer: 203450-08-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5432e4e37f42cf05fbc4f03ce9941c583d17d777&vid=5432e4e37f42cf05fbc4f03ce9941c583d17d777&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 231.0 to 235.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR