N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-piperidone ≥98.0%

Leverantör: TCI EUROPE

Synonyms: N-Fmoc-4-piperidone

F1031-1G F1031-5G
TCIAF1031-1GEA 575 SEK
TCIAF1031-1G TCIAF1031-5G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-piperidone ≥98.0%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-piperidone
Formel: C₂₀H₁₉NO₃
MW: 321,38 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00673794
CAS nummer: 204376-55-6

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 96.0 %
Melting point 131.0 to 135.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR