3-Fluoro-5-(trifluoromethyl)benzonitrile ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
F0994-5G
F0994-25G
TCIAF0994-5GEA
1175
SEK
TCIAF0994-5G
TCIAF0994-25G
3-Fluoro-5-(trifluoromethyl)benzonitrile ≥98.0%
3-Fluoro-5-(trifluoromethyl)benzonitrile
Formel:
C₈H₃F₄N MW: 189,11 g/mol Flampunkt: 65°C |
MDL Number:
MFCD00061282 CAS nummer: 149793-69-1 UN: 3276 ADR: 6.1,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...