4-Fluoro-3-(trifluoromethyl)benzonitrile ≥98.0%
Leverantör: TCI
|
Fara
|
Synonyms:
5-Cyano-2-fluorobenzotrifluoride
F0979-5G
F0979-25G
TCIAF0979-5GEA
455
SEK
TCIAF0979-5G
TCIAF0979-25G
4-Fluoro-3-(trifluoromethyl)benzonitrile ≥98.0%
4-Fluoro-3-(trifluoromethyl)benzonitrile
Formel:
C₈H₃F₄N |
MDL Number:
MFCD00061284 CAS nummer: 67515-59-7 UN: 3439 ADR: 6.1,III |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 66.0 to 70.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...