5-Fluoro-2-methylphenylboronic acid (contains varying amounts of Anhydride)

Leverantör: TCI EUROPE
Varning

Synonyms: 5-Fluoro-2-methylbenzeneboronic acid, 2-Methyl-5-fluorophenylboronic acid

F0976-1G F0976-5G
TCIAF0976-1GEA 295 SEK
TCIAF0976-1G TCIAF0976-5G
5-Fluoro-2-methylphenylboronic acid (contains varying amounts of Anhydride)
5-Fluoro-2-methylphenylboronic acid
Formel: C₇H₈BFO₂
MW: 153,95 g/mol
MDL Number: MFCD03095047
CAS nummer: 163517-62-2

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 114.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR