4-Fluorobenzenesulphonamide ≥98.0%
Leverantör: TCI
|
Varning
|
F0953-5G
F0953-25G
TCIAF0953-5GEA
725
SEK
TCIAF0953-5G
TCIAF0953-25G
4-Fluorobenzenesulphonamide ≥98.0%
4-Fluorobenzenesulphonamide
Formel:
C₆H₆FNO₂S |
MDL Number:
MFCD00025384 CAS nummer: 402-46-0 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 125.0 to 129.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...