2-Propanesulphonyl chloride ≥97%
Leverantör: Thermo Scientific
Fara
Synonyms: Isopropylsulphonyl chloride
L02431.06 L02431.14
L02431.06EA 1100 SEK
L02431.06 L02431.14
2-Propanesulphonyl chloride ≥97%
2-Propanesulphonyl chloride
Purity: 97% CAS No.: 10147-37-2 Molecular Formula: (CH3)2CHSO2Cl Formula Weight: 142.60 Flash Point: 86°(186°F) Refractive Index: 1.4560 Melting Point: -47° Boiling Point: 95°/45mm Beilstein Reference: 1747497 UN No.: 3265 MDL No.: MFCD00007453
Formel: C₃H₇ClO₂S
MW: 142,61 g/mol
Kokpunkt: 95 °C (45 mmHg)
Smältpunkt: –47 °C
Densitet: 1,273
Flampunkt: 86 °C (186 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00007453
CAS nummer: 10147-37-2
EINECS: 233-415-2
UN: 3265
ADR: 8,II