(R)-N-Fmoc-(2-furyl)alanine ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: Fmoc-beta-(2-furyl)-D-Ala-OH, N-Fmoc-3-(2-furyl)-D-alanine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(2-furyl)-D-alanine

F0934-200MG F0934-1G
TCIAF0934-200MGEA 765 SEK
TCIAF0934-200MG TCIAF0934-1G
(R)-N-Fmoc-(2-furyl)alanine ≥98.0% (by HPLC, titration analysis)
(R)-N-Fmoc-(2-furyl)alanine
Formel: C₂₂H₁₉NO₅
MW: 377,396 g/mol
MDL Number: MFCD01311774
CAS nummer: 220497-85-8

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 146.0 to 150.0 deg-C
Specific rotation [a]20/D +33.0 to +37.0 deg (C=1,DMF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR