Thiazole-5-carboxyaldehyde ≥98.0%

Leverantör: TCI
Varning

Synonyms: 5-Thiazolecarboxaldehyde, 5-Formylthiazole

F0908-1G F0908-5G
TCIAF0908-1GEA 870 SEK
TCIAF0908-1G TCIAF0908-5G
Thiazole-5-carboxyaldehyde ≥98.0%
Thiazole-5-carboxyaldehyde
Formel: C₄H₃NOS
MW: 113.14 g/mol
Kokpunkt: 94°C
Flampunkt: 98°C
Storage Temperature: Kylskåp
MDL Number: MFCD02179516
CAS nummer: 1003-32-3

Order Now

Specifikation testresultat

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR