4-Fluoro-D-2-phenylglycine ≥98.0%

Leverantör: TCI EUROPE

Synonyms: H-D-Phg(4-F)-OH, (R)-2-Amino-2-(4-fluorophenyl)acetic Acid

F0863-1G F0863-5G
TCIAF0863-1GEA 975 SEK
TCIAF0863-1G TCIAF0863-5G
4-Fluoro-D-2-phenylglycine ≥98.0%
4-Fluoro-D-2-phenylglycine
Formel: C₈H₈FNO₂
MW: 169,16 g/mol
MDL Number: MFCD00042727
CAS nummer: 93939-74-3
EINECS: 300-363-8

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -130.0 to -140.0 deg (C=1, 1mol/L HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR