2-Fluoro-5-(trifluoromethyl)benzonitrile ≥98.0%
Leverantör: TCI EUROPE
Fara
Synonyms: 2-Fluoro-5-trifluoromethylbenzonitrile, 3-Cyano-4-fluorobenzotrifluoride
F0860-1G F0860-5G
TCIAF0860-1GEA 335 SEK
TCIAF0860-1G TCIAF0860-5G
2-Fluoro-5-(trifluoromethyl)benzonitrile ≥98.0%
2-Fluoro-5-(trifluoromethyl)benzonitrile
Formel: C₈H₃F₄N
MW: 189,11 g/mol
Kokpunkt: 99°C
MDL Number: MFCD00061280
CAS nummer: 4088-84-0
UN: 3276
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %